missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Boc-amino)ethylamino]-3-iodopyridine, 95%, Alfa Aesar™

Product Code. 11444520
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11444520

Brand: Thermo Scientific Alfa Aesar H50092.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 902835-89-6
Molecular Formula C12H18IN3O2
Molecular Weight (g/mol) 363.199
MDL Number MFCD07781137
InChI Key RHBATGHZRRPELS-UHFFFAOYSA-N
Synonym tert-butyl n-2-3-iodopyridin-2-yl amino ethyl carbamate, tert-butyl 2-3-iodopyridin-2-ylamino ethyl carbamate, 2-2-boc-amino ethylamino-3-iodopyridine, tert-butyl 2-3-iodopyridin-2-yl amino ethyl carbamate, tert-butoxy-n-2-3-iodo 2-pyridyl amino ethyl carboxamide
PubChem CID 28054477
IUPAC Name tert-butyl N-[2-[(3-iodopyridin-2-yl)amino]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCNC1=C(C=CC=N1)I

Specifications

Quantity 1 g
Formula Weight 363.2
Percent Purity 95%
Chemical Name or Material 2-[2-(Boc-amino)ethylamino]-3-iodopyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.