Learn More
2-(2-Methoxyphenoxy)ethylamine, 97%, Thermo Scientific™
78.10 EUR - 310.00 EUR
Chemical Identifiers
CAS | 1836-62-0 |
---|---|
Molecular Formula | C9H13NO2 |
Molecular Weight (g/mol) | 167.208 |
MDL Number | MFCD00235185 |
InChI Key | CKJRKLKVCHMWLV-UHFFFAOYSA-N |
Synonym | 2-2-methoxyphenoxy ethylamine, 2-2-methoxyphenoxy ethanamine, 1-2-aminoethoxy-2-methoxybenzene, 2-2-methoxyphenoxy ethyl amine, 2-2-methoxy-phenoxy-ethylamine, ethanamine, 2-2-methoxyphenoxy, unii-20s0vx4qt9, 2-2'-methoxy phenoxyethylamine, 2-2-methoxyphenoxy ethan-1-amine |
PubChem CID | 1713005 |
IUPAC Name | 2-(2-methoxyphenoxy)ethanamine |
SMILES | COC1=CC=CC=C1OCCN |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2-(2-Methoxyphenoxy)ethylamine is used for its reaction with 4-(2,3-epoxypropoxy) carbazole for preparation of Carvedilol (I).
Solubility
Difficult to mix.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
1836-62-0 | |
167.208 | |
CKJRKLKVCHMWLV-UHFFFAOYSA-N | |
1713005 | |
COC1=CC=CC=C1OCCN |
C9H13NO2 | |
MFCD00235185 | |
2-2-methoxyphenoxy ethylamine, 2-2-methoxyphenoxy ethanamine, 1-2-aminoethoxy-2-methoxybenzene, 2-2-methoxyphenoxy ethyl amine, 2-2-methoxy-phenoxy-ethylamine, ethanamine, 2-2-methoxyphenoxy, unii-20s0vx4qt9, 2-2'-methoxy phenoxyethylamine, 2-2-methoxyphenoxy ethan-1-amine | |
2-(2-methoxyphenoxy)ethanamine |
Specifications
1836-62-0 | |
98°C (0.4 mmHg) | |
C9H13NO2 | |
UN2735 | |
2-2-methoxyphenoxy ethylamine, 2-2-methoxyphenoxy ethanamine, 1-2-aminoethoxy-2-methoxybenzene, 2-2-methoxyphenoxy ethyl amine, 2-2-methoxy-phenoxy-ethylamine, ethanamine, 2-2-methoxyphenoxy, unii-20s0vx4qt9, 2-2'-methoxy phenoxyethylamine, 2-2-methoxyphenoxy ethan-1-amine | |
CKJRKLKVCHMWLV-UHFFFAOYSA-N | |
2-(2-methoxyphenoxy)ethanamine | |
1713005 | |
97% |
1.11 | |
Amine-like | |
MFCD00235185 | |
Air Sensitive | |
Difficult to mix. | |
COC1=CC=CC=C1OCCN | |
167.208 | |
167.21 | |
2-(2-Methoxyphenoxy)ethylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.