missing translation for 'onlineSavingsMsg'
Learn More
Alfa Aesar™ 2,3,6-Trifluoropyridine-4-carboxylic acid, tech. 90% Product Code.: 11324476

Alfa Aesar™ 2,3,6-Trifluoropyridine-4-carboxylic acid, tech. 90%

Product Code. 11324476
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11324476

Brand: Alfa Aesar™ L20051.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 675602-92-3
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.082
MDL Number MFCD03840204
InChI Key RXNORRUCMWSIQZ-UHFFFAOYSA-N
Synonym 2,3,6-trifluoroisonicotinic acid, 2, 3, 6-trifluoroisonicotinic acid, 2,3,6-trifluoro-4-pyridinecarboxylic acid, 4-pyridinecarboxylicacid,2,3,6-trifluoro, 4-pyridinecarboxylic acid, 2,3,6-trifluoro, 2,3,6-tris fluoranyl pyridine-4-carboxylic acid, 2,3,6-trifluoropyridine-4-carboxylic acid
PubChem CID 2782837
IUPAC Name 2,3,6-trifluoropyridine-4-carboxylic acid
SMILES C1=C(C(=C(N=C1F)F)F)C(=O)O

Specifications

Melting Point 118°C to 121°C
Quantity 250 mg
Formula Weight 177.08
Percent Purity 90%
Grade Technical
Chemical Name or Material 2,3,6-Trifluoropyridine-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alfa Aesar™ 2,3,6-Trifluoropyridine-4-carboxylic acid, tech. 90% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.