missing translation for 'onlineSavingsMsg'
Learn More

2,3-dimethyl-1h-indole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-4510300
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10738825

Brand: Thermo Scientific Maybridge MO07746CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 14844-73-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
InChI Key KHJGIMZYCBPOBG-UHFFFAOYSA-N
PubChem CID 693386
IUPAC Name 2,3-dimethyl-1H-indole-5-carboxylic acid
SMILES CC1=C(NC2=C1C=C(C=C2)C(=O)O)C

Specifications

Assay Percent Range 97%
Quantity 250 mg
Synonym 1h-indole-5-carboxylic acid, 2,3-dimethyl, 2,3-dimethylindole-5-carboxylic acid, 5-carboxy-2,3-dimethyl-1h-indole, 2,3-dimethyl-indole-5-carboxylic acid, 1h-indole-5-carboxylicacid,2,3-dimethyl, 1h-indole-5-carboxylicacid, 2,3-dimethyl, 2,3-dimethyl-1h-indole-5-carboxylic acid
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethyl-1H-indole-5-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.