Learn More
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%, Thermo Scientific Chemicals
88.24 EUR - 297.48 EUR
Chemical Identifiers
CAS | 191171-55-8 |
---|---|
Molecular Formula | C12H18BNO2 |
Molecular Weight (g/mol) | 219.09 |
MDL Number | MFCD02179448 |
InChI Key | ZCJRWQDZPIIYLM-UHFFFAOYSA-N |
Synonym | 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-aminophenylboronic acid pinacol ester, 2-aminophenylboronic acid, pinacol ester, 2-aminophenylboronicacidpinacolester, 2-aminophenylboronic acid pinacol cyclic ester, 2-aminobenzeneboronic acid pinacol ester, 2-aminophenyl boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl |
PubChem CID | 2734652 |
IUPAC Name | 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=CC=C1N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10662803
|
Thermo Scientific Acros
367560010 |
1 g | Glass bottle |
88.24 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10232572
|
Thermo Scientific Acros
367560050 |
5 g | Glass bottle |
297.48 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
191171-55-8 | |
219.09 | |
ZCJRWQDZPIIYLM-UHFFFAOYSA-N | |
2734652 | |
CC1(C)OB(OC1(C)C)C1=CC=CC=C1N |
C12H18BNO2 | |
MFCD02179448 | |
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-aminophenylboronic acid pinacol ester, 2-aminophenylboronic acid, pinacol ester, 2-aminophenylboronicacidpinacolester, 2-aminophenylboronic acid pinacol cyclic ester, 2-aminobenzeneboronic acid pinacol ester, 2-aminophenyl boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
Specifications
191171-55-8 | |
100.0 | |
Cream | |
96% min. (GC) | |
MFCD02179448 | |
Solubility in water: insoluble in water. | |
CC1(C)OB(OC1(C)C)C1=CC=CC=C1N | |
219.09 | |
219.09 | |
Crystalline Powder |
96.0 | |
64.0°C to 70.0°C | |
Authentic | |
C12H18BNO2 | |
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-aminophenylboronic acid pinacol ester, 2-aminophenylboronic acid, pinacol ester, 2-aminophenylboronicacidpinacolester, 2-aminophenylboronic acid pinacol cyclic ester, 2-aminobenzeneboronic acid pinacol ester, 2-aminophenyl boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
ZCJRWQDZPIIYLM-UHFFFAOYSA-N | |
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
2734652 | |
97% | |
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
Recommended Storage : product may turn dark on exposure to air
- 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.