missing translation for 'onlineSavingsMsg'
Learn More

2,4-Thiazolidinedione, 99%

Product Code. 10113310 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10113310

Brand: Thermo Scientific Acros 159491000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2295-31-0 | C3H3NO2S | 117.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2295-31-0
Molecular Formula C3H3NO2S
Molecular Weight (g/mol) 117.12
MDL Number MFCD00005478
InChI Key ZOBPZXTWZATXDG-UHFFFAOYSA-N
Synonym 2,4-thiazolidinedione, thiazolidinedione, thiazolidine-2,4-dione, 2,4-dioxothiazolidine, 2,4 3h,5h-thiazoledione, usaf ek-5496, thiazolidindione, unii-aa68lxk93c, 2,4 thiazolidinedione, thiazolidinedione-2,4
PubChem CID 5437
ChEBI CHEBI:50992
IUPAC Name 1,3-thiazolidine-2,4-dione
SMILES C1C(=O)NC(=O)S1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 123°C to 126°C
Color Yellow
Boiling Point 178°C to 179°C (19.0 mmHg)
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 27, 233
Formula Weight 117.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Thiazolidinedione
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Thiazolidinedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.