missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichlorothiophene-3-carbonyl chloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 215.49 |
Percent Purity | 95% |
Chemical Name or Material | 2, 5-Dichlorothiophene-3-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'