missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,5-Dimethoxythiophenol, 95%, Thermo Scientific™

Product Code. p-7045700
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11392537

Brand: Alfa Aesar™ L11641.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1483-27-8
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD00052738
InChI Key SESUUAOAUZDHHP-UHFFFAOYSA-N
Synonym 2,5-dimethoxythiophenol, 2,5-dimethoxybenzene-1-thiol, benzenethiol, 2,5-dimethoxy, pubchem6852, acmc-1cgdz, 3,6-dimethoxybenzenethiol, #, 2,5-dimethoxyphenyl mercaptan
PubChem CID 519009
SMILES COC1=CC(S)=C(OC)C=C1

Specifications

Density 1.14
Boiling Point 278°C
Flash Point 122°C (252°F)
Odor Stench
Refractive Index 1.5845
Quantity 1 g
UN Number UN3334
Beilstein 1867842
Sensitivity Air Sensitive
IUPAC Name 2,5-dimethoxybenzenethiol
Formula Weight 170.23
Percent Purity 95%
Chemical Name or Material 2,5-Dimethoxythiophenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.