missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-pyrroline, 75%, mixture of cis and trans, Thermo Scientific™

Product Code. p-119472 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10104470

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 59480-92-1
Molecular Formula C6H12N
Molecular Weight (g/mol) 98.17
MDL Number MFCD00005215
InChI Key TXQDHQBSNAJSHQ-OLQVQODUSA-O
PubChem CID 101066
IUPAC Name 2,5-dimethyl-2,5-dihydro-1H-pyrrole
SMILES C[C@H]1[NH2+][C@@H](C)C=C1

Specifications

CAS Min % 70.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8240g/mL
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Refractive Index 1.4404
Quantity 1 g
Specific Gravity 0.824
Synonym 2,5-dimethyl-3-pyrroline, 2,5-dimethyl-3-pyrroline, mixture of cis and trans, 2,5-dimethyl-2,5-dihydropyrrole, 1h-pyrrole,2,5-dihydro-2,5-dimethyl, 1h-pyrrole, 2,5-dihydro-2,5-dimethyl
Formula Weight 97.16
Percent Purity 75%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethyl-3-pyrroline, 75%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethyl-3-pyrroline, mixture of cis and trans
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.