missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromocarbazole

Product Code. 17221236 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17221236

Brand: Thermo Scientific Acros 467780010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136630-39-2 | C12H7Br2N | 325.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136630-39-2
Molecular Formula C12H7Br2N
Molecular Weight (g/mol) 325.00
InChI Key QPTWWBLGJZWRAV-UHFFFAOYSA-N
IUPAC Name 2,7-dibromo-9H-carbazole
SMILES BrC1=CC2=C(C=C1)C1=C(N2)C=C(Br)C=C1

Specifications

Melting Point 225.0°C to 230.0°C
Quantity 1 g
Formula Weight 325
Chemical Name or Material 2,7-Dibromocarbazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.