missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-difluorobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11349965
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83506-93-8
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.12
MDL Number MFCD00077509
InChI Key DGOZIZVTANAGCA-UHFFFAOYSA-N
Synonym 4,5-difluoroanthranilic acid, 2-amino-4,5-difluoro-benzoic acid, buttpark 49\07-41, benzoicacid, 2-amino-4,5-difluoro, benzoic acid, 2-amino-4,5-difluoro, pubchem4675, 2-amino-4,5-difluoro, 4,5-difluoroantranilic acid, ksc448a3l, 4,5-difluoro anthranilic acid
PubChem CID 735965
IUPAC Name 2-amino-4,5-difluorobenzoic acid
SMILES NC1=CC(F)=C(F)C=C1C(O)=O

Specifications

Melting Point 181°C to 183°C
Quantity 5 g
Beilstein 2834162
Formula Weight 173.12
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5-difluorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'