Learn More
2-Amino-6-fluorobenzothiazole, 99%, Thermo Scientific™
21.01 EUR
Chemical Identifiers
CAS | 348-40-3 |
---|---|
MDL Number | MFCD00013336 |
InChI Key | CJLUXPZQUXVJNF-UHFFFAOYSA-N |
Synonym | 2-amino-6-fluorobenzothiazole, 6-fluorobenzo d thiazol-2-amine, 6-fluoro-benzothiazol-2-ylamine, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 6-fluorobenzothiazole-2-ylamine, pubchem14467, maybridge1_006546 |
PubChem CID | 319954 |
IUPAC Name | 6-fluoro-1,3-benzothiazol-2-amine |
SMILES | C1=CC2=C(C=C1F)SC(=N2)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10590394
|
Thermo Scientific Acros
263730010 |
1 g | Glass bottle | ||||||
Description
Chemical Identifiers
348-40-3 | |
CJLUXPZQUXVJNF-UHFFFAOYSA-N | |
319954 | |
C1=CC2=C(C=C1F)SC(=N2)N |
MFCD00013336 | |
2-amino-6-fluorobenzothiazole, 6-fluorobenzo d thiazol-2-amine, 6-fluoro-benzothiazol-2-ylamine, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 6-fluorobenzothiazole-2-ylamine, pubchem14467, maybridge1_006546 | |
6-fluoro-1,3-benzothiazol-2-amine |
Specifications
348-40-3 | |
100.0 | |
Tan to White | |
98.5% min. (GC) | |
MFCD00013336 | |
CJLUXPZQUXVJNF-UHFFFAOYSA-N | |
6-fluoro-1,3-benzothiazol-2-amine | |
319954 | |
99% | |
2-Amino-6-fluorobenzothiazole, 99% |
98.5 | |
181.0°C to 185.0°C | |
Authentic | |
C7H5FN2S | |
2-amino-6-fluorobenzothiazole, 6-fluorobenzo d thiazol-2-amine, 6-fluoro-benzothiazol-2-ylamine, 2-benzothiazolamine, 6-fluoro, 6-fluoro-2-benzothiazolamine, 2-amino-6-fluoro benzothiazole, 2-amino-6-fluoro-1,3-benzothiazole, 6-fluorobenzothiazole-2-ylamine, pubchem14467, maybridge1_006546 | |
C1=CC2=C(C=C1F)SC(=N2)N | |
168.2 | |
168.2 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'tsca' : TSCA
- 2-Amino-6-fluorobenzothiazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only