missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric acid, 99%, Thermo Scientific Chemicals

Product Code. 11399679
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-57-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008049
InChI Key FUOOLUPWFVMBKG-UHFFFAOYSA-N
Synonym 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
PubChem CID 6119
ChEBI CHEBI:27971
IUPAC Name 2-amino-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)N

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 506496
Merck Index 14,445
Solubility Information Soluble in water. Insoluble in alcohol and ether.
Formula Weight 103.12
Percent Purity 99%
Chemical Name or Material 2-Aminoisobutyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'