missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)benzeneboronic acid, 96%, Thermo Scientific™

Product Code. p-7034749
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15499187

Brand: Thermo Scientific Alfa Aesar H52448.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115377-94-1
Molecular Formula C11H16BNO4
Molecular Weight (g/mol) 237.062
MDL Number MFCD02179450
InChI Key RMEIKYSECODAHJ-UHFFFAOYSA-N
Synonym 2-boc-aminophenyl boronic acid, 2-tert-butoxycarbonyl amino phenyl boronic acid, n-tert-butoxycarbonyl-2-amino-1-phenylboronic acid, 2-n-boc-amino phenylboronic acid, 2-tert-butoxycarbonylamino phenylboronic acid, n-tert-butoxycarbonyl 2-amino-1-phenylboronic acid, 2-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 2-tert-butoxycarbonyl amino phenylboronic acid, 2-t-butoxycarbonylamino phenylboronic acid, carbamic acid, n-2-boronophenyl-, 1,1-dimethylethyl ester
PubChem CID 2773226
IUPAC Name [2-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
SMILES B(C1=CC=CC=C1NC(=O)OC(C)(C)C)(O)O

Specifications

Melting Point 123°C to 125°C
Quantity 250 mg
Formula Weight 237.06
Percent Purity 96%
Chemical Name or Material 2-(Boc-amino)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.