Learn More
2-Bromo-3-nitrobenzoic acid, tech. 90%, Thermo Scientific Chemicals
22.30 EUR - 66.13 EUR
Chemical Identifiers
CAS | 573-54-6 |
---|---|
Molecular Formula | C7H4BrNO4 |
Molecular Weight (g/mol) | 246.016 |
MDL Number | MFCD00074899 |
InChI Key | WTDJEGSXLFHZPY-UHFFFAOYSA-N |
Synonym | 2-bromo-3-nitrobenzoic acid, benzoic acid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoicacid, 2-bromo-3-nitro-benzoic acid, pubchem4714, acmc-1an8y, ksc273m5t, benzoicacid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoic acid, technical gradeShow More |
PubChem CID | 68452 |
SMILES | C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O |
Description
2-Bromo-3-nitrobenzoic acid is used in preparation of 3-substituted 5-nitroisocoumarins1 and 2-bromo-3-nitrobenzaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-3-nitrobenzoic acid is used in preparation of 3-substituted 5-nitroisocoumarins1 and 2-bromo-3-nitrobenzaldehyde.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.
Chemical Identifiers
573-54-6 | |
246.016 | |
WTDJEGSXLFHZPY-UHFFFAOYSA-N | |
68452 |
C7H4BrNO4 | |
MFCD00074899 | |
2-bromo-3-nitrobenzoic acid, benzoic acid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoicacid, 2-bromo-3-nitro-benzoic acid, pubchem4714, acmc-1an8y, ksc273m5t, benzoicacid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoic acid, technical gradeShow More | |
C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O |
Specifications
573-54-6 | |
C7H4BrNO4 | |
2-bromo-3-nitrobenzoic acid, benzoic acid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoicacid, 2-bromo-3-nitro-benzoic acid, pubchem4714, acmc-1an8y, ksc273m5t, benzoicacid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoic acid, technical grade | |
C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O | |
68452 | |
90% | |
2-Bromo-3-nitrobenzoic acid |
184°C to 186°C | |
MFCD00074899 | |
WTDJEGSXLFHZPY-UHFFFAOYSA-N | |
246.016 | |
246.02 | |
Technical |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 209-356-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.