missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorothiophene, 97%, Thermo Scientific Chemicals

Product Code. 11492668
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2873-18-9
Molecular Formula C4H2BrClS
Molecular Weight (g/mol) 197.47
MDL Number MFCD00014523
InChI Key ZFAJPWYXLYGUJU-UHFFFAOYSA-N
Synonym 2-chloro-5-bromothiophene, 5-bromo-2-chlorothiophene, 2-chloro-5-bromo thiophene, thiophene, 2-bromo-5-chloro, 2-brom-5-chlorthiophen, pubchem5279, acmc-1cs3j, 2-bromo-5-chloro-thiophene, 5-bromo-2-chloro-thiophene, ksc207m3t
PubChem CID 76133
IUPAC Name 2-bromo-5-chlorothiophene
SMILES ClC1=CC=C(Br)S1

Specifications

Density 1.803
Boiling Point 69°C to 70°C (18 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.596
Quantity 5 g
UN Number UN2810
Beilstein 109897
Formula Weight 197.48
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-chlorothiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'