missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, Honeywell
71.21 EUR - 829.04 EUR
Chemical Identifiers
CAS | 78-93-3 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00011648 |
InChI Key | ZWEHNKRNPOVVGH-UHFFFAOYSA-N |
Synonym | 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon |
PubChem CID | 6569 |
ChEBI | CHEBI:28398 |
IUPAC Name | butan-2-one |
SMILES | CCC(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15623950
|
Honeywell Riedel-de Haën
33407-1L |
1 L |
71.21 EUR
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15633950
|
Honeywell Riedel-de Haën
33407-2.5L |
2.5 L |
159.84 EUR
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15653950
|
Honeywell Riedel-de Haën
33407-30KG |
30 kg | N/A | ||||||
Description
Chemical Identifiers
78-93-3 | |
72.11 | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
6569 | |
butan-2-one |
C4H8O | |
MFCD00011648 | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
CHEBI:28398 | |
CCC(C)=O |
Specifications
78-93-3 | |
0.101 | |
960°F | |
C2H5COCH3 | |
71 mmHg (20°C) | |
741880 | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
butan-2-one | |
6569 | |
72.11g/mol | |
2-Butanone |
0.805g/mL at 25°C | |
−3°C (26.6°F) | |
C4H8O | |
MFCD00011648 | |
UN1193 | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
CCC(C)=O | |
72.11 | |
CHEBI:28398 | |
≥99.5% (GC) |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
Danger
missing translation for 'einecsNumber' : 201-159-0
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Butanone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title