missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Chloro-4-(trifluoromethyl)pyrimidine, 97%
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Boiling Point | 60°C (10 mmHg) |
Flash Point | 49°C (120°F) |
Quantity | 5 g |
UN Number | UN1993 |
Formula Weight | 182.53 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-4-(trifluoromethyl)pyrimidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'