missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(methoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15440286
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Boronic acids are often used in metal-catalyzed cross-coupling for C-C, C-N bond formation, and C-H functionalization. The widely utilized Suzuki-Miyaura coupling allows for the coupling of boronic acids with carbon halides and surrogates. Other known chemistry may includes 1,4-addition to activated alkenes, 1,2-addition to carbonyls and imines, addition to anhydrides, 1,4-addition to a,ß-unsaturated ketones(4), and 1,4-addition to esters and amides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-92-4
Molecular Formula C8H8BClO4
Molecular Weight (g/mol) 214.408
MDL Number MFCD08689528
InChI Key LZMDHZSRUQKDFI-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxycarbonyl phenylboronic acid, methyl 3-borono-4-chlorobenzoate, 2-chloro-5-methoxycarbonyl phenyl boronic acid, 2-chloro-5-methoxycarbonyl phenylboronicacid, acmc-209rad, 2-chloro-5-methoxycarbonylphenyl boronic acid, 2-chloranyl-5-methoxycarbonyl-phenyl boronic acid, benzoicacid, 3-borono-4-chloro-, 1-methyl ester 9ci
PubChem CID 44119360
IUPAC Name (2-chloro-5-methoxycarbonylphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)OC)Cl)(O)O

Specifications

Melting Point 230°C to 232°C
Quantity 1 g
Formula Weight 214.41
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-5-(methoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.