missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-cyclopentyl-4-nitrobenzamide, 97%, Thermo Scientific™

191.00 EUR - 615.00 EUR

Chemical Identifiers

CAS 300829-22-5
Molecular Formula C12H13ClN2O3
Molecular Weight (g/mol) 268.70
MDL Number MFCD00707289
InChI Key SWXOJSLQXXNFQE-UHFFFAOYSA-N
Synonym cambridge id 5346233, 2-chloro-4-nitrophenyl-n-cyclopentylcarboxamide
PubChem CID 710872
IUPAC Name 2-chloro-N-cyclopentyl-4-nitrobenzamide
SMILES [O-][N+](=O)C1=CC(Cl)=C(C=C1)C(=O)NC1CCCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15485398
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
H59168.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15465398
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
H59168.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

300829-22-5
268.70
SWXOJSLQXXNFQE-UHFFFAOYSA-N
710872
[O-][N+](=O)C1=CC(Cl)=C(C=C1)C(=O)NC1CCCC1
C12H13ClN2O3
MFCD00707289
cambridge id 5346233, 2-chloro-4-nitrophenyl-n-cyclopentylcarboxamide
2-chloro-N-cyclopentyl-4-nitrobenzamide

Specifications

300829-22-5
MFCD00707289
SWXOJSLQXXNFQE-UHFFFAOYSA-N
2-chloro-N-cyclopentyl-4-nitrobenzamide
710872
97%
C12H13ClN2O3
cambridge id 5346233, 2-chloro-4-nitrophenyl-n-cyclopentylcarboxamide
[O-][N+](=O)C1=CC(Cl)=C(C=C1)C(=O)NC1CCCC1
268.70
268.7
2-Chloro-N-cyclopentyl-4-nitrobenzamide
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.