Learn More
MP Biomedicals™ 2% Cholesterol Diet, Pellet
Product Code. 11452751 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 kg
20 kg
50 kg
Unit Size:
10kg
20kg
50kg
Product Code. 11452751
Brand: MP Biomedicals™ 0290469110
Complete rat feed with 2% cholesterol added.
  • 2% Cholesterol
  • For rodent diet
  • Pellets
TRUSTED_SUSTAINABILITY
Specifications
Product Type Animal Diet
For Use With (Application) Rodent diet
Quantity 10 kg
Storage Requirements 2°C to 8°C