missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | White to Yellow |
Density | 1.0610g/mL |
Flash Point | 110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | [(CH3)2CH]2NP(Cl)O(CH2)2CN |
Refractive Index | 1.4760 to 1.4800 |
Show More |
Safety and Handling
Product Identifier
- 2-Cyanoethyl N,N-diisopropylchlorophosphoramidite
- Danger
- Serious eye damage/eye irritation Category 1
- Pyrophoric liquid Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H250-Catches fire spontaneously if exposed to air.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction