missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanothioacetamide, 97%

Product Code. p-3757342 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10217040

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7357-70-2 | C3H4N2S | 100.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7357-70-2
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00010025
InChI Key BHPYMZQTCPRLNR-UHFFFAOYSA-N
Synonym 2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide
PubChem CID 1416277
IUPAC Name 2-cyanoethanethioamide
SMILES NC(=S)CC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 113°C to 119°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NCCH2CSNH2
Quantity 5 g
Formula Weight 100.14
Percent Purity 97%
Physical Form Needles or Powder
Chemical Name or Material 2-Cyanothioacetamide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.