missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Diphenylphosphino)benzaldehyde oxime, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Quantity | 1 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 305.32 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-(Diphenylphosphino)benzaldehyde oxime |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction