missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoro-4-hydroxybenzoic acid, 98%, Thermo Scientific™ Product Code.: 15545256

2-Fluoro-4-hydroxybenzoic acid, 98%, Thermo Scientific™

Product Code. 15545256
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15545256

Brand: Thermo Scientific Acros 457590050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65145-13-3
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
MDL Number MFCD01310985
InChI Key NXWTWYULZRDBSA-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid, benzoic acid, 2-fluoro-4-hydroxy, pubchem2608, acmc-1b9jp, 4-hydroxyfluorobenzoic acid, frinton fr-2445, ksc352q5p, 2-fluoro-4-hydroxybenzoicacid, 4-hydroxy-2-fluorobenzoic acid, 2-fluoro4-hydroxy-benzoic acidShow MoreShow Less
PubChem CID 2783398
IUPAC Name 2-fluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1

Specifications

Melting Point 187.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-hydroxybenzoic acid
compliance-icons
Product Identifier
  • 2-Fluoro-4-hydroxybenzoic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoro-4-hydroxybenzoic acid, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.