Learn More
2-Fluoro-4-nitrotoluene, 98%
Description
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.
Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 32°C to 37°C |
| Boiling Point | 56°C to 58°C (1 mmHg) |
| Flash Point | 73°C (163°F) |
| Quantity | 25 g |
| UN Number | UN2811 |
| Beilstein | 2250156 |
| Solubility Information | Insoluble in water. Solubility in methanol gives very faint turbidity. |
| Formula Weight | 155.13 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Fluoro-4-nitrotoluene |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.