Learn More
2-Hexene, cis + trans, tech. 85%, Thermo Scientific Chemicals
78.89 EUR - 289.44 EUR
Chemical Identifiers
CAS | 592-43-8 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.162 |
MDL Number | MFCD00009473 |
InChI Key | RYPKRALMXUUNKS-HWKANZROSA-N |
Synonym | cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z |
PubChem CID | 639661 |
IUPAC Name | (E)-hex-2-ene |
SMILES | CCCC=CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15420328
|
Thermo Scientific Alfa Aesar
H53472.06 |
5 g |
78.89 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15430328
|
Thermo Scientific Alfa Aesar
H53472.14 |
25 g |
289.44 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
592-43-8 | |
84.162 | |
RYPKRALMXUUNKS-HWKANZROSA-N | |
639661 | |
CCCC=CC |
C6H12 | |
MFCD00009473 | |
cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z | |
(E)-hex-2-ene |
Specifications
592-43-8 | |
67°C to 69°C | |
C6H12 | |
1.395 | |
UN3295 | |
cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z | |
CCCC=CC | |
84.162 | |
84.16 | |
Technical |
0.678 | |
−20°C (−4°F) | |
CH3(CH2)2CH=CHCH3 | |
MFCD00009473 | |
969179 | |
RYPKRALMXUUNKS-HWKANZROSA-N | |
(E)-hex-2-ene | |
639661 | |
85% | |
2-Hexene, Cis + Trans |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 209-755-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.