missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinobenzoic acid hydrochloride, 99%

Product Code. p-143563 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10038941

Brand: Thermo Scientific Acros 411800050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52356-01-1 | C7H9ClN2O2 | 188.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52356-01-1
Molecular Formula C7H9ClN2O2
Molecular Weight (g/mol) 188.61
MDL Number MFCD00012931
InChI Key ZGNNOFKURIXXRF-UHFFFAOYSA-N
Synonym 2-hydrazinobenzoic acid hydrochloride, 2-hydrazinylbenzoic acid hydrochloride, benzoic acid, 2-hydrazino-, monohydrochloride, benzoic acid, 2-hydrazino-, hydrochloride, benzoic acid, 2-hydrazinyl-, hydrochloride 1:1, 2-carboxyphenylhydrazine hydrochloride, 2-hydrazinobenzoicacidhydrochloride, 2-carboxyphenylhydrazinium 1+ chloride, pubchem6411, 2-hydrazinobenzoic acid hydrochloride 1:1
PubChem CID 104169
IUPAC Name 2-hydrazinylbenzoic acid;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 175°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 188.61
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Hydrazinobenzoic acid hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydrazinobenzoic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.