missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-(trifluoromethyl)pyridine, 98%, Thermo Scientific™

Product Code. 11319979
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11319979

Brand: Thermo Scientific Alfa Aesar H25756.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Hydroxy-4-(trifluoromethyl)pyridine is used in the production of agrochemicals and pharmaceuticals.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50650-59-4
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.10
MDL Number MFCD00153195,MFCD09029339
InChI Key IKHLLNMSMFVTLP-UHFFFAOYSA-N
Synonym 2-hydroxy-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-2-ol, 4-trifluoromethyl pyridin-2 1h-one, 4-trifluoromethyl-pyridin-2-ol, 4-trifluoromethyl-2-pyridone, 2-hydroxy-4-trifluoromethylpyridine, 4-trifluoromethyl-2 1h pyridinone, 4-trifluoromethyl-1h-pyridin-2-one, 2 1h-pyridinone, 4-trifluoromethyl, 4-trifluoromethyl-2 1h-pyridinone
PubChem CID 2775130
SMILES FC(F)(F)C1=CC(=O)NC=C1

Specifications

Melting Point 158°C to 160°C
Quantity 250 mg
Solubility Information Soluble in methanol.
IUPAC Name 4-(trifluoromethyl)-1H-pyridin-2-one
Formula Weight 163.1
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-4-(trifluoromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.