missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1H-imidazole-4-carbaldehyde, 97%, Thermo Scientific™

45.10 EUR - 680.00 EUR

Chemical Identifiers

CAS 35034-22-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
InChI Key ZWULFIBGPXWGFG-UHFFFAOYSA-N
Synonym 2-methyl-1h-imidazole-4-carbaldehyde, 2-methyl-4-formylimidazole, 2-methylimidazole-4-carbaldehyde, 4-formyl-2-methyl-1h-imidazole, 2-methylimidazole-5-carboxaldehyde, 2-methyl-3h-imidazole-4-carbaldehyde, 2-methyl-1h-imidazole-4-carboxaldehyde, 4-formyl-2-methylimidazole, 2-methyl-4-imidazolecarboxaldehyde, 2-methyl-1h-imidazole-5-carboxaldehyde
PubChem CID 2795878
IUPAC Name 2-methyl-1H-imidazole-5-carbaldehyde
SMILES CC1=NC=C(N1)C=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10693413
View Documents
Thermo Scientific Maybridge
SB01361DA
1 g Amber Glass Bottle
45.10 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
10192934
View Documents
Thermo Scientific Maybridge
SB01361EA
10 g Amber Glass Bottle
349.00 EUR
10g
Please to purchase this item. Need a web account? Register with us today!
10549434
View Documents
Thermo Scientific Maybridge
SB01361EB
25 g Amber Glass Bottle
680.00 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

35034-22-1
110.116
2-methyl-1h-imidazole-4-carbaldehyde, 2-methyl-4-formylimidazole, 2-methylimidazole-4-carbaldehyde, 4-formyl-2-methyl-1h-imidazole, 2-methylimidazole-5-carboxaldehyde, 2-methyl-3h-imidazole-4-carbaldehyde, 2-methyl-1h-imidazole-4-carboxaldehyde, 4-formyl-2-methylimidazole, 2-methyl-4-imidazolecarboxaldehyde, 2-methyl-1h-imidazole-5-carboxaldehyde
2-methyl-1H-imidazole-5-carbaldehyde
C5H6N2O
ZWULFIBGPXWGFG-UHFFFAOYSA-N
2795878
CC1=NC=C(N1)C=O

Specifications

35034-22-1
C5H6N2O
ZWULFIBGPXWGFG-UHFFFAOYSA-N
2-methyl-1H-imidazole-5-carbaldehyde
2795878
2-Methyl-1H-imidazole-4-carbaldehyde
97%
2-methyl-1h-imidazole-4-carbaldehyde, 2-methyl-4-formylimidazole, 2-methylimidazole-4-carbaldehyde, 4-formyl-2-methyl-1h-imidazole, 2-methylimidazole-5-carboxaldehyde, 2-methyl-3h-imidazole-4-carbaldehyde, 2-methyl-1h-imidazole-4-carboxaldehyde, 4-formyl-2-methylimidazole, 2-methyl-4-imidazolecarboxaldehyde, 2-methyl-1h-imidazole-5-carboxaldehyde
CC1=NC=C(N1)C=O
110.116
110.12
Videos
SDS
Documents

Documents

Product Certifications