missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-hexanone, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Specifications
Specifications
CAS | 7379-12-6 |
Density | 0.8250g/mL |
Boiling Point | 131.0°C to 132.0°C |
Flash Point | 24°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Molecular Formula | C7H14O |
Quantity | 5 g |
Specific Gravity | 0.825 |
Synonym | 2-methyl-3-hexanone, 3-hexanone, 2-methyl, isopropyl propyl ketone, 2-methyl-hexan-3-one, propyl isopropyl ketone, isopropyl-n-propyl ketone, 4-01-00-03328 beilstein handbook reference |
Show More |
Safety and Handling
Product Identifier
- 2-Methyl-3-hexanone, 98%
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction