missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Methylacetophenone, 98%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
2'-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.
Solubility
Soluble in ethanol. Insoluble in water.
Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2′-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.
Solubility
Soluble in ethanol. Insoluble in water.
Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 1.02 |
| Boiling Point | 214°C |
| Flash Point | 75°C (167°F) |
| Refractive Index | 1.5318 |
| Quantity | 5 g |
| Beilstein | 907005 |
| Solubility Information | Soluble in ethanol. Insoluble in water. |
| Formula Weight | 134.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 2'-Methylacetophenone |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction