Learn More
2-Octen-4-one, 97%, Thermo Scientific Chemicals
63.11 EUR - 241.20 EUR
Chemical Identifiers
CAS | 4643-27-0 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.199 |
MDL Number | MFCD00061023 |
InChI Key | FMDLEUPBHMCPQV-GQCTYLIASA-N |
Synonym | 2-octen-4-one, propenyl butyl ketone, butyl propenyl ketone, unii-c9nb51lmxt, 2-octen-4-one natural, fema no. 3603, c9nb51lmxt, e-oct-2-en-4-one, 2e-oct-2-en-4-one, 2e-2-octen-4-one |
PubChem CID | 5365891 |
IUPAC Name | (E)-oct-2-en-4-one |
SMILES | CCCCC(=O)C=CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11405906
|
Thermo Scientific Alfa Aesar
B22250.03 |
1 g |
63.11 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415906
|
Thermo Scientific Alfa Aesar
B22250.06 |
5 g |
241.20 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
4643-27-0 | |
126.199 | |
FMDLEUPBHMCPQV-GQCTYLIASA-N | |
5365891 | |
CCCCC(=O)C=CC |
C8H14O | |
MFCD00061023 | |
2-octen-4-one, propenyl butyl ketone, butyl propenyl ketone, unii-c9nb51lmxt, 2-octen-4-one natural, fema no. 3603, c9nb51lmxt, e-oct-2-en-4-one, 2e-oct-2-en-4-one, 2e-2-octen-4-one | |
(E)-oct-2-en-4-one |
Specifications
4643-27-0 | |
65°C (15 mmHg) | |
C8H14O | |
MFCD00061023 | |
2-octen-4-one, propenyl butyl ketone, butyl propenyl ketone, unii-c9nb51lmxt, 2-octen-4-one natural, fema no. 3603, c9nb51lmxt, e-oct-2-en-4-one, 2e-oct-2-en-4-one, 2e-2-octen-4-one | |
FMDLEUPBHMCPQV-GQCTYLIASA-N | |
(E)-oct-2-en-4-one | |
5365891 | |
97% |
0.85 | |
58°C (136°F) | |
1.445 | |
UN1224 | |
Insoluble in water. | |
CCCCC(=O)C=CC | |
126.199 | |
126.2 | |
2-Octen-4-one |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 225-071-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.