missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-propanol, 99%, Thermo Scientific Chemicals

27.44 EUR - 238.19 EUR

Chemical Identifiers

CAS 617-94-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004456
InChI Key BDCFWIDZNLCTMF-UHFFFAOYSA-N
Synonym 2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
PubChem CID 12053
IUPAC Name 2-phenylpropan-2-ol
SMILES CC(C)(O)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10735611
SDS View Documents
Thermo Scientific Acros
130830050
5 g Glass bottle
27.44 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
10133280
SDS View Documents
Thermo Scientific Acros
130830250
25 g Glass bottle
73.87 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
10475485
SDS View Documents
Thermo Scientific Acros
130831000
100 g Glass bottle
238.19 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

617-94-7
136.19
BDCFWIDZNLCTMF-UHFFFAOYSA-N
12053
CC(C)(O)C1=CC=CC=C1
C9H12O
MFCD00004456
2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
2-phenylpropan-2-ol

Specifications

617-94-7
100.0
Colorless to Yellow
88°C to 90°C (11.0 mmHg)
Authentic
C9H12O
1.5190 to 1.522
06, 506
2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
BDCFWIDZNLCTMF-UHFFFAOYSA-N
2-phenylpropan-2-ol
12053
99%
2-Phenyl-2-propanol
98.5
32°C to 34°C
0.9730g/mL
88°C
98.5% min. (GC)
C6H5C(CH3)2OH
MFCD00004456
0.973
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,benzene
CC(C)(O)C1=CC=CC=C1
136.19
136.19
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 210-539-5

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.