Learn More
Ismatec™ 2-Stop Tubing, Tygon HC F-4040-A
Click to view available options
Tubing Size:
I.D. 0.38 mm
I.D. 1.42 mm
I.D. 1.52 mm
Unit Size:
Pack of 12
Description
- Specially formulated to transport hydrocarbons, petroleum products and distillates.
- Ideal for gasoline, kerosene, heating oils, cutting liquids and coolants based on glycols.
- Low gas permeability.
Specifications
Specifications
| Color Code | Yellow-Yellow |
| Diameter (Metric) Inner | 1.42 mm |
| Material | Tygon HC |
| Overall Length (Metric) | 400 mm |
| For Use With (Application) | Pumps that incorporate cassettes into the pumphead design. Hydrocarbons, petroleum products, and distillates. |
| Min. Operating Temperature (Metric) | −37°C |
| Packaging Quantity | 12/Pk |
| Resistance Properties | Acids: Limited, Alkaline solutions: Not rec., Solvents: Not rec., Pressure: Good, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited |
| Tubing Size | I.D. 1.42 mm |
| Color | Amber |
| Show More |