missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific™

Product Code. 15224278
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 154630-93-0
Molecular Formula C6H10F3N
Molecular Weight (g/mol) 153.15
InChI Key NAXDEFXCCITWEU-UHFFFAOYSA-N
Synonym 2-trifluoromethyl piperidine, 2-trifluoromethyl-piperidine, -2-trifluoromethyl piperidine, piperidine, 2-trifluoromethyl, +/--2-trifluoromethyl piperidine
PubChem CID 11263608
IUPAC Name 2-(trifluoromethyl)piperidine
SMILES C1CCNC(C1)C(F)(F)F

Specifications

Boiling Point 64.0°C (182.0 mmHg)
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 153.15
Percent Purity 97%
Chemical Name or Material (±)-2-(Trifluoromethyl)piperidine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (±)-2-(Trifluoromethyl)piperidine, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'