Learn More
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh
Shop All Fisherbrand Products
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
1.00 mm
106 μm
1.18 mm
125 μm
1.40 mm
150 μm
1.70 mm
180 μm
2 mm
212 μm
2.36 mm
250 μm
2.80 mm
300 μm
3.35 mm
355 μm
425 μm
500 μm
600 μm
710 μm
852 μm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1 Each |
| Pore Size | 45 μm |
| Type | Sieve |
| Certifications/Compliance | Part 1 : 2000, ISO 3310 : Part 1 : 2000, BS410 |
| Material | Stainless Steel |
| Includes | Certificate of conformity and record card |
| Diameter (Metric) Outer | 200 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Woven Wire |