Learn More
BRAND™ 24-well Insert 2in1, PC-Membrane
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
Brand:  BRAND™ 782707
 View more versions of this product
Product Code. 16194947
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Use Inserts in a hanging or standing position
  • For 24-well plates or culture dishes
  • Achieve uniform and controlled cell growth due to surface treatment, without coatings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24-well Insert 2in1, PC-Membrane 8 μm, Multipack
8 μm
Sterile
Polycarbonate
4 x 12
24