Learn More
3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, 98%, Thermo Scientific Chemicals
53.37 EUR - 231.15 EUR
Chemical Identifiers
CAS | 175136-66-0 |
---|---|
Molecular Formula | C17H6F12O |
Molecular Weight (g/mol) | 454.215 |
MDL Number | MFCD00042474 |
InChI Key | GATWMPGNBWPCIY-UHFFFAOYSA-N |
Synonym | 3,3',5,5'-tetrakis trifluoromethyl benzophenone, bis 3,5-bis trifluoromethyl phenyl methanone, maybridge1_002937, acmc-1c7yy, bis 3,5-bis trifluoromethyl phenyl ketone, di 3,5-di trifluoromethyl phenyl methanone, bis 3,5-bis trifluoromethyl phenyl methanone #, 3,3/',5,5/'-tetrakis trifluoromethyl benzophenoneShow More |
PubChem CID | 519439 |
IUPAC Name | bis[3,5-bis(trifluoromethyl)phenyl]methanone |
SMILES | C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479093
|
Thermo Scientific Alfa Aesar
A11164.03 |
1 g |
53.37 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489093
|
Thermo Scientific Alfa Aesar
A11164.06 |
5 g |
231.15 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,3′,5,5′-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.
Solubility
Sparingly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
175136-66-0 | |
454.215 | |
GATWMPGNBWPCIY-UHFFFAOYSA-N | |
519439 | |
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F |
C17H6F12O | |
MFCD00042474 | |
3,3',5,5'-tetrakis trifluoromethyl benzophenone, bis 3,5-bis trifluoromethyl phenyl methanone, maybridge1_002937, acmc-1c7yy, bis 3,5-bis trifluoromethyl phenyl ketone, di 3,5-di trifluoromethyl phenyl methanone, bis 3,5-bis trifluoromethyl phenyl methanone #, 3,3/',5,5/'-tetrakis trifluoromethyl benzophenoneShow More | |
bis[3,5-bis(trifluoromethyl)phenyl]methanone |
Specifications
175136-66-0 | |
C17H6F12O | |
3,3',5,5'-tetrakis trifluoromethyl benzophenone, bis 3,5-bis trifluoromethyl phenyl methanone, maybridge1_002937, acmc-1c7yy, bis 3,5-bis trifluoromethyl phenyl ketone, di 3,5-di trifluoromethyl phenyl methanone, bis 3,5-bis trifluoromethyl phenyl methanone #, 3,3/',5,5/'-tetrakis trifluoromethyl benzophenone | |
GATWMPGNBWPCIY-UHFFFAOYSA-N | |
bis[3,5-bis(trifluoromethyl)phenyl]methanone | |
519439 | |
98% |
139°C to 140°C | |
MFCD00042474 | |
Sparingly soluble in water. | |
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F | |
454.215 | |
454.22 | |
3,3',5,5'-Tetrakis (trifluoromethyl)benzophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.