Learn More
3,4-Dimethylhexane, 97%, Thermo Scientific Chemicals
96.78 EUR - 370.85 EUR
Chemical Identifiers
CAS | 583-48-2 |
---|---|
Molecular Formula | C8H18 |
Molecular Weight (g/mol) | 114.232 |
MDL Number | MFCD00039927 |
InChI Key | RNTWWGNZUXGTAX-UHFFFAOYSA-N |
Synonym | hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso |
PubChem CID | 11412 |
IUPAC Name | 3,4-dimethylhexane |
SMILES | CCC(C)C(C)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15430388
|
Thermo Scientific Alfa Aesar
H53498.06 |
5 g |
96.78 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15440388
|
Thermo Scientific Alfa Aesar
H53498.14 |
25 g |
370.85 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Chemical Identifiers
583-48-2 | |
114.232 | |
RNTWWGNZUXGTAX-UHFFFAOYSA-N | |
11412 | |
CCC(C)C(C)CC |
C8H18 | |
MFCD00039927 | |
hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso | |
3,4-dimethylhexane |
Specifications
583-48-2 | |
119°C | |
C8H18 | |
MFCD00039927 | |
1718744 | |
RNTWWGNZUXGTAX-UHFFFAOYSA-N | |
3,4-dimethylhexane | |
11412 | |
97% |
0.72 | |
6°C (43°F) | |
1.403 | |
UN1262 | |
hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso | |
CCC(C)C(C)CC | |
114.232 | |
114.23 | |
3,4-Dimethylhexane |
Safety and Handling
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
missing translation for 'einecsNumber' : 209-504-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.