Learn More
3,5-Dichlorophenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Product Code. 11439157
Click to view available options
Quantity:
5 g
25 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6590-93-8
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00041072
InChI Key JLEMKZDHFGCHLO-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl isothiocyanate, 3,5-dichlorophenylisothiocyanate, 1,3-dichloro-5-isothiocyanato-benzene, benzene,1,3-dichloro-5-isothiocyanato, benzene, 1,3-dichloro-5-isothiocyanato, 3,5-dichlorobenzenisothiocyanate, acmc-20and6, 3,5-dichloroisothiocyanatobenzene, #
PubChem CID 138776
IUPAC Name 1,3-dichloro-5-isothiocyanatobenzene
SMILES ClC1=CC(=CC(Cl)=C1)N=C=S
Specifications
Melting Point 44°C to 48°C
Boiling Point 137°C to 138°C (9 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 25 g
UN Number UN2923
Beilstein 879282
Sensitivity Moisture sensitive
Formula Weight 204.08
Percent Purity 98%
Chemical Name or Material 3,5-Dichlorophenyl isothiocyanate