missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-hydroxybenzonitrile, 97%, Thermo Scientific Chemicals

Product Code. 15404227
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2967-54-6
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD09033181
InChI Key XZNZJDPPWWFJAL-UHFFFAOYSA-N
Synonym 3,5-difluoro-4-hydroxy-benzonitrile, 2,6-difluoro-4-cyanophenol, 4-cyano-2,6-difluorophenol, 3,5-difluoro-4-hydroxy benzonitrile, pubchem23504, benzonitrile, 3,5-difluoro-4-hydroxy
PubChem CID 10749406
IUPAC Name 3,5-difluoro-4-hydroxybenzonitrile
SMILES OC1=C(F)C=C(C=C1F)C#N

Specifications

Melting Point 200°C to 208°C
Quantity 250 mg
UN Number UN3439
Formula Weight 155.1
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-4-hydroxybenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'