missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpiperidine, cis + trans, 97%, Thermo Scientific Chemicals

Product Code. 11405417
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35794-11-7
Molecular Formula C7H16N
Molecular Weight (g/mol) 114.21
MDL Number MFCD00005996,MFCD09832871
InChI Key IDWRJRPUIXRFRX-KNVOCYPGSA-O
Synonym piperidine, 3,5-dimethyl, 3,5-lupetidine, 3,5-dimethylpiperidin, 3,5-dimethylpiperidine, cis + trans, pubchem7709, 3,5-dimethylpiperdine, 3,5 dimethylpiperidine, 3,5-dimethylpiperadine, acmc-1afvq, hexahydro-3,5-lutidine
PubChem CID 118259
IUPAC Name 3,5-dimethylpiperidine
SMILES C[C@H]1C[NH2+]C[C@@H](C)C1

Specifications

Density 0.83
Boiling Point 142°C to 146°C
Flash Point 32°C (89°F)
Refractive Index 1.4454
Quantity 100 g
UN Number UN1993
Beilstein 102638
Formula Weight 113.2
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylpiperidine, cis + trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'