Learn More
3-Amino-2-chloropyridine, 98+%
Description
It is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.
Solubility
Soluble in methanol and water. (30 g/L).
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 76°C to 80°C |
| Boiling Point | 130°C to 134°C (22 mmHg) |
| Flash Point | 185°C (365°F) |
| Odor | Unpleasant |
| Quantity | 50 g |
| Beilstein | 109814 |
| Solubility Information | Soluble in methanol and water. (30g/L). |
| Formula Weight | 128.56 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 3-Amino-2-chloropyridine |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.