missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole, 98+%, Thermo Scientific Chemicals

Product Code. 11382526
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3314-35-0
Molecular Formula C9H11N3
Molecular Weight (g/mol) 161.208
MDL Number MFCD00051730
InChI Key QENUTIJJGGTTPE-UHFFFAOYSA-N
Synonym 1-phenyl-4,5-dihydro-1h-pyrazol-3-amine, 2-pyrazoline, 3-amino-1-phenyl, 1-fenyl-3-aminopyrazolin, 3-amino-1-phenyl-2-pyrazoline, 1-phenyl-4,5-dihydro-1h-pyrazol-3-ylamine, 3-amino-4,5-dihydro-1-phenylpyrazole, pyrazolidine, 3-imino-1-phenyl, 4,5-dihydro-1-phenyl-1h-pyrazol-3-amine, 3-amino-1-fenyl-2-pyrazolin, 1-phenyl-3-amino-2-pyrazoline
PubChem CID 98736
IUPAC Name 2-phenyl-3,4-dihydropyrazol-5-amine
SMILES C1CN(N=C1N)C2=CC=CC=C2

Specifications

Melting Point 167°C to 170°C
Quantity 2 g
Beilstein 139162
Formula Weight 161.21
Percent Purity ≥98%
Chemical Name or Material 3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'