missing translation for 'onlineSavingsMsg'
Learn More
3-Aminooxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals Product Code.: 15443798

3-Aminooxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15443798
500 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15443798

Brand: Thermo Scientific Alfa Aesar H57291.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138650-24-5
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD11035888
InChI Key LXRNYRAHXDUNKR-UHFFFAOYSA-N
Synonym 3-oxetanecarboxylic acid, 3-amino, 3-amino-oxetane-3-carboxylic acid, acmc-20a7wi, 3-amino-3-oxetanecarboxylic acid, 3-aminooxetane-3-carboxylic acidShow MoreShow Less
PubChem CID 15024256
IUPAC Name 3-aminooxetane-3-carboxylic acid
SMILES C1C(CO1)(C(=O)O)N

Specifications

Melting Point 205°C to 210°C
Color White
Quantity 500 mg
Formula Weight 117.1
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Aminooxetane-3-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ24G023 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57291.MD,H57291.ME
Lot NumberG17Y008 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57291.MD,H57291.ME
Lot NumberP09H036 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57291.MD,H57291.ME

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Aminooxetane-3-carboxylic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.