missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-butanone, 97%, stab. with <1% magnesium oxide, Thermo Scientific Chemicals

154.77 EUR - 1169.82 EUR

Chemical Identifiers

CAS 814-75-5
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151.003
MDL Number MFCD00013538
InChI Key BNBOUFHCTIFWHN-UHFFFAOYSA-N
Synonym 3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on
PubChem CID 13142
IUPAC Name 3-bromobutan-2-one
SMILES CC(C(=O)C)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11419492
View Documents
Thermo Scientific Alfa Aesar
A10186.09
10 g
154.77 EUR
10g
Please to purchase this item. Need a web account? Register with us today!
11429492
View Documents
Thermo Scientific Alfa Aesar
A10186.18
50 g
528.63 EUR
50g
Please to purchase this item. Need a web account? Register with us today!
11439492
View Documents
Thermo Scientific Alfa Aesar
A10186.30
250 g
1169.82 EUR
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3-Bromo-2-butanone is involved in the preparation of o-isopropyl S-3-oxobutan-2-yl dithiocarbonate by reaction with potassium o-isopropylxanthate, which on treatment with sulfuric acid gives 4,5-dimethyl-1,3-dithiol-2-one .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-butanone is involved in the preparation of o-isopropyl S-3-oxobutan-2-yl dithiocarbonate by reaction with potassium o-isopropylxanthate, which on treatment with sulfuric acid gives 4,5-dimethyl-1,3-dithiol-2-one .

Solubility
Miscible with dichloromethane.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications

Chemical Identifiers

814-75-5
151.003
BNBOUFHCTIFWHN-UHFFFAOYSA-N
13142
CC(C(=O)C)Br
C4H7BrO
MFCD00013538
3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on
3-bromobutan-2-one

Specifications

814-75-5
69°C to 70°C (70 mmHg)
Pungent
1.459
UN2922
3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on
BNBOUFHCTIFWHN-UHFFFAOYSA-N
3-bromobutan-2-one
13142
97%
1.438
>100°C (212°F)
C4H7BrO
MFCD00013538
906773
Miscible with dichloromethane.
CC(C(=O)C)Br
151.003
151.01
3-Bromo-2-butanone, Stabilized with <1% magnesium oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H314-H318-H312
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302+H312+H332-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 212-404-6

missing translation for 'rtecsNumber' : EL7005000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.