missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxybenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11493997
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2315-86-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00143096
InChI Key HLHNOIAOWQFNGW-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r
PubChem CID 75341
IUPAC Name 3-bromo-4-hydroxybenzonitrile
SMILES OC1=CC=C(C=C1Br)C#N

Specifications

Melting Point 156°C to 161°C
Quantity 100 g
UN Number UN3439
Beilstein 2207020
Formula Weight 198.02
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-hydroxybenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'