missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxypyridine, 97%

Product Code. 15431429
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15431429

Brand: Thermo Scientific Alfa Aesar H64296.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82257-09-8 | C6H6BrNO | 188.024 g/mol

3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water (9.8 g/L).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82257-09-8
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD05664024
InChI Key DSFJKFHIMBVWAV-UHFFFAOYSA-N
PubChem CID 642827
IUPAC Name 3-bromo-4-methoxypyridine
SMILES COC1=C(C=NC=C1)Br

Specifications

Color Yellow
Quantity 1 g
Beilstein 1423610
Synonym 3-bromo-4-methoxy-pyridine, pyridine, 3-bromo-4-methoxy, pyridine,3-bromo-4-methoxy, pubchem6686, 4-methoxy-3-bromopyridine, acmc-209po0, abbypharma ap-17-5958
Solubility Information Slightly soluble in water (9.8g/L).
Formula Weight 188.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methoxypyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.