missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorophenol, 97%, Thermo Scientific Chemicals

Product Code. 15461498
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433939-27-6
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD07783710
InChI Key JCPJGUPQZDEZQH-UHFFFAOYSA-N
Synonym 3-fluoro-5-bromophenol, 5-bromo-3-fluorophenol, 3-bromo-5-fluoro-phenol, phenol, 3-bromo-5-fluoro, pubchem4129, acmc-1apwb, ksc493m6t, 3-bromo-5-fluorophenol
PubChem CID 21904636
IUPAC Name 3-bromo-5-fluorophenol
SMILES C1=C(C=C(C=C1F)Br)O

Specifications

Melting Point 36°C to 40°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-fluorophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'